Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPN12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527733 Supplier Invitrogen™ Supplier No. PA527733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2. Predicted reactivity: Mouse (99%), Rat (98%), Xenopus laevis (81%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PTP PEST is an 88 kDa cytosolic protein tyrosine phosphatase which is ubiquitiously expressed in mammalian tissues. PTP PEST exhibits a high specific activity when assayed in vitro with artificial tyrosine-phosphorylated substrates. PTP PEST is subject to regulation via phosphorylation of ser 39 and is catalyzed by both protein kinase C and protein kinase A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN12
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PTPN12
Gene Accession No. Q05209
Gene Alias MPTP-PEST; P19-PTP; pest; protein tyrosine phosphatase non-receptor type 12; protein tyrosine phosphatase, non-receptor type 12; protein-tyrosine phosphatase G1; Protein-tyrosine phosphatase P19; PTPG1; PTPN12; PTP-P19; PTP-PEST; RKPTP; tcag7.1075; Tyrosine-protein phosphatase non-receptor type 12
Gene Symbols PTPN12
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human PTPN12. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117255, 5782
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.