Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984347
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4347 Supplier Bioss Supplier No. BS19589R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTP PEST is an 88 kDa cytosolic protein tyrosine phosphatase which is ubiquitiously expressed in mammalian tissues. PTP PEST exhibits a high specific activity when assayed in vitro with artificial tyrosine-phosphorylated substrates. PTP PEST is subject to regulation via phosphorylation of ser 39 and is catalyzed by both protein kinase C and protein kinase A.

Specifications

Antigen PTPN12
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PTPN12
Gene Accession No. Q05209
Gene Alias BOS_4175; MPTP-PEST; P19-PTP; protein tyrosine phosphatase, non-receptor type 12; Protein-tyrosine phosphatase G1; Protein-tyrosine phosphatase P19; PTPG1; Ptpn12; PTP-P19; PTP-PEST; RKPTP; tcag7.1075; Tyrosine-protein phosphatase non-receptor type 12
Gene Symbols PTPN12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PTPN12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5782
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.