Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPN12 Recombinant Rabbit Monoclonal Antibody (BL-5-2F8)

Catalog No. PIMA544210
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544210 20 μL
PIMA544209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544210 Supplier Invitrogen™ Supplier No. MA544210
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

PTP PEST is an 88 kDa cytosolic protein tyrosine phosphatase which is ubiquitiously expressed in mammalian tissues. PTP PEST exhibits a high specific activity when assayed in vitro with artificial tyrosine-phosphorylated substrates. PTP PEST is subject to regulation via phosphorylation of ser 39 and is catalyzed by both protein kinase C and protein kinase A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN12
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BL-5-2F8
Concentration 40 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6
Gene PTPN12
Gene Accession No. Q05209
Gene Alias MPTP-PEST; P19-PTP; pest; protein tyrosine phosphatase non-receptor type 12; protein tyrosine phosphatase, non-receptor type 12; protein-tyrosine phosphatase G1; Protein-tyrosine phosphatase P19; PTPG1; PTPN12; PTP-P19; PTP-PEST; RKPTP; tcag7.1075; Tyrosine-protein phosphatase non-receptor type 12
Gene Symbols PTPN12
Host Species Rabbit
Immunogen Synthetic peptide representing a region between residues 499 and 780 of human Protein Tyrosine Phosphatase, non-receptor type 12.
Purification Method Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5782
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.