Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPN13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572907
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572907 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572907 Supplier Invitrogen™ Supplier No. PA572907
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTPN13 Polyclonal Antibody for Western Blot, IHC (P), IP

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP is a large intracellular protein. It has a catalytic PTP domain at its C-terminus and two major structural domains: a region with five PDZ domains and a FERM domain that binds to plasma membrane and cytoskeletal elements. This PTP was found to interact with, and dephosphorylate, Fas receptor and IkappaBalpha through the PDZ domains. This suggests it has a role in Fas mediated programmed cell death. This PTP was also shown to interact with GTPase-activating protein, and thus may function as a regulator of Rho signaling pathways. Four alternatively spliced transcript variants, which encode distinct proteins, have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN13
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.02% sodium azide
Gene PTPN13
Gene Accession No. Q12923
Gene Alias AI324989; DKFZp686J1497; FAP-1; Fas-associated protein-tyrosine phosphatase 1; hPTP1E; hPTPE1; PNP1; protein Tyr phosphatase; protein tyrosine phosphatase DPZPTP; Protein tyrosine phosphatase PTP-BL; protein tyrosine phosphatase, non-receptor type 13; protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associated phosphatase); protein-tyrosine phosphatase 1E; Protein-tyrosine phosphatase PTPL1; protein-tyrosine phosphatase RIP; ptp; Ptp14; PTP1E; PTP36; PTP-BAS; PTP-BL; PTP-E1; PTPL1; PTPLE; PTPN13; Ptpri; RIP; tyrosine phosphatase; tyrosine-protein phosphatase non-receptor type 13
Gene Symbols PTPN13
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1760-1775 of human FAP-1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5783
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.