Learn More
Description
Specifications
Specifications
| Antigen | PTPN13/PTPL1 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000 |
| Formulation | Whole antisera with 0.02% Sodium Azide |
| Gene Accession No. | Q12923 |
| Gene Alias | hPTPE1, PNP1, protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associatedphosphatase), protein-tyrosine phosphatase 1, Fas-associated, PTP1EDKFZp686J1497, PTP-BASEC 3.1.3.48, PTP-E1, PTPL1nonreceptor type 13, tyrosine-protein phosphatase non-receptor type 13 |
| Gene Symbols | PTPN13 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
