Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPN14 Mouse anti-Human, Clone:2B3H7, Abnova™
Mouse monoclonal antibody raised against recombinant human PTPN14.
Supplier: Abnova Corporation MAB17310
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal noncatalytic domain similar to that of band 4.1 superfamily cytoskeleton-associated proteins, which suggested the membrane or cytoskeleton localization of this protein. The specific function of this PTP has not yet been determined. [provided by RefSeq]Specifications
PTPN14 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-400) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1000) Immunofluorescence (1:200-1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by th |
|
PTPN14 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2B3H7 | |
protein tyrosine phosphatase, non-receptor type 14 | |
In PBS (0.05% sodium azide) | |
MGC126803, PEZ, PTP36 | |
Recombinant protein corresponding to amino acids 896-1169 of of human PTPN14 from E. coli. | |
5784 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction