Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN14 Mouse anti-Human, Clone:2C12E3, Abnova™

Catalog No. 89333568 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-568 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-568 Supplier Abnova Corporation Supplier No. MAB17311
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human PTPN14.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal noncatalytic domain similar to that of band 4.1 superfamily cytoskeleton-associated proteins, which suggested the membrane or cytoskeleton localization of this protein. The specific function of this PTP has not yet been determined. [provided by RefSeq]

Specifications

Antigen PTPN14
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2C12E3
Conjugate Unconjugated
Description protein tyrosine phosphatase, non-receptor type 14
Dilution ELISA (1:10000)
Flow Cytometry (1:200-400)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1000)
Immunofluorescence (1:200-1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by th
Formulation In PBS (0.05% sodium azide)
Gene PTPN14
Gene Alias MGC126803, PEZ, PTP36
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 896-1169 of of human PTPN14 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 5784
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.