Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPN18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584151
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584151 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584151 Supplier Invitrogen™ Supplier No. PA584151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEAPLYSKVT PRAQRPGAHA EDARGTLPGR VPADQSPAGS GAYEDVAGGA QTGGLGFNLR IGRPK Highest antigen sequence indentity to the following orthologs: Rat - 65%, Mouse - 68%.

Differentially dephosphorylate autophosphorylated tyrosine kinases which are known to be overexpressed in tumor tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN18
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPN18
Gene Accession No. Q99952
Gene Alias BDP1; Brain-derived phosphatase; fetal liver phosphatase 1; FLP1; FLP-1; HSCF; protein tyrosine phosphatase 20; protein tyrosine phosphatase, non-receptor type 18; protein tyrosine phosphatase, non-receptor type 18 (brain-derived); PTP20; PTP-HSCF; Ptpk1; PTP-K1; Ptpn18; Tyrosine-protein phosphatase non-receptor type 18
Gene Symbols PTPN18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTPN18. Recombinant protein control fragment (Product #RP-108398).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26469
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.