Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPN18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113316
Description
PTPN18 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Differentially dephosphorylate autophosphorylated tyrosine kinases which are known to be overexpressed in tumor tissues.
Specifications
PTPN18 | |
Polyclonal | |
Unconjugated | |
PTPN18 | |
BDP1; Brain-derived phosphatase; fetal liver phosphatase 1; FLP1; FLP-1; HSCF; protein tyrosine phosphatase 20; protein tyrosine phosphatase, non-receptor type 18; protein tyrosine phosphatase, non-receptor type 18 (brain-derived); PTP20; PTP-HSCF; Ptpk1; PTP-K1; Ptpn18; Tyrosine-protein phosphatase non-receptor type 18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26469, 301333 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99952 | |
PTPN18 | |
Recombinant Human Tyrosine-protein phosphatase non-receptor type 18 protein (1-210AA). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction