Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582516 Supplier Invitrogen™ Supplier No. PA582516
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDINIKQVLL NMRKYRMGLI QTPDQLRFSY MAIIEGAKCI KGDSSIQKRW KELSKEDLSP AFDHSPNKIM TEKYNGNRIG LEEEKLTGDR CTGLSSKMQD TMEENSESAL RKRIREDRKA TTAQKVQQMK QRLNENERK Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 68%.

T-cell protein tyrosine phosphatase (TC-PTP), also known as PTPT and PTPN2, is an enzyme that removes phosphate groups covalently attached to tyrosine residues in proteins. This enzyme has two C-terminal end splice variants with distinctly different subcellular localizations. The shorter 45 kilodalton isoform is exclusively nuclear in resting cells, but redistrubutes to the cytosol upon stimulation with growth factors and cellular stress. The longer 48 kilodalton isoform is exclusively found in the endoplasmic reticulum 3 and seems to have distinctly different physiologic substrates from the smaller isoform. Although found in many cell types and tissues, TC-PTP is particularly prominent in hemopoietic cell types. Knockout mice lacking TC-PTP are born viable but die 3 to 5 weeks after birth of erythropoietic and lymphopoietic deficits 7, indicating a critical role for TC-PTP in bone marrow maturation. TC-PTP will dephosphorylate a wide range of phosphoproteins, such as p52 Shc 6 and receptors for EGF 1, Insulin 8 and growth hormone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPN2
Gene Accession No. P17706
Gene Alias AI325124; hypothetical protein LOC540888; MPTP; protein tyrosine phosphatase non-receptor type 2; protein tyrosine phosphatase, non-receptor type 2; protein-tyrosine phosphatase PTP-2; Protein-tyrosine phosphatase PTP-S; PTN2; PTPN 2; PTPN2; Ptpt; T cell protein-tyrosine phosphatase; TC PTP; T-cell protein tyrosine phosphatase; T-cell protein-tyrosine phosphatase; TCELLPTP; TCPTP; TC-PTP; tyrosine-phosphatase; Tyrosine-protein phosphatase non-receptor type 2
Gene Symbols PTPN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTPN2. Recombinant protein control fragment (Product #RP-90507).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5771
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.