Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN2/TCPTP Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569754
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9754 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9754 Supplier Bethyl Laboratories, Inc Supplier No. A305185AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat

T cell protein tyrosine phosphatase (TCPTP), also known as PTPT and PTPN2, is an enzyme that removes phosphate groups covalently attached to tyrosine residues in proteins. TCPTP is ubiquitously expressed with particularly high expression in hematopoietic tissues and exists as two forms: an endoplasmic reticulum-targeted 48-kDa form (TC48) and a nuclear 45-kDa form (TC45). Although targeted to the nucleus, TC45 can exit in response to specific stimuli to dephosphorylate cytoplasmic substrates. TC-PTP is a negative regulator of the Jak/Stat cytokine signaling pathway. TCPTP is the PTP most closely related to PTP1B, with 72% sequence identity and 86% similarity in the catalytic domain.

Specifications

Antigen PTPN2/TCPTP
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PTPN2
Gene Accession No. P17706
Gene Alias PTPN2
Gene Symbols PTPN2
Host Species Rabbit
Immunogen Between 350 and 400
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5771
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.