Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPN20A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560332 Supplier Invitrogen™ Supplier No. PA560332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NFPHNDYEDV FEEPSESGSD PSMWTARG Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 39%.

Tyrosine-protein phosphatase targeted to sites of actin polymerization in response of varied extracellular stimuli. Has tyrosine phosphatase activity towards various tyrosyl phosphorylated substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN20A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPN20
Gene Accession No. Q4JDL3
Gene Alias bA142I17.1; bA42B19.1; CT126; hPTPN20; protein tyrosine phosphatase, non-receptor type 20; protein tyrosine phosphatase, non-receptor type 20A; protein tyrosine phosphatase, non-receptor type 20B; PTPN20; PTPN20A; PTPN20B; Tyrosine-protein phosphatase non-receptor type 20
Gene Symbols PTPN20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTPN20A. Recombinant protein control fragment (Product #RP-104358).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26095
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.