Learn More
Invitrogen™ PTPN22 Polyclonal Antibody

Description
The antibody detects endogenous levels of total PTPN22 protein.
Specifications
Specifications
| Antigen | PTPN22 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Ptpn22 |
| Gene Accession No. | P29352, Q9Y2R2 |
| Gene Alias | 70zpep; Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP; Lymphoid phosphatase; lymphoid-specific protein tyrosine phosphatase; LyP; LYP1; LYP2; PEP; PEST domain-enriched tyrosine phosphatase; PEST-domain phosphatase; protein tyrosine phosphatase, non-receptor type 22; protein tyrosine phosphatase, non-receptor type 22 (lymphoid); protein tyrosine phosphatase, non-receptor type 8; Ptpn22; PTPN8; tyrosine-phosphatase; Tyrosine-protein phosphatase non-receptor type 22 |
| Gene Symbols | Ptpn22 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.