Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN23 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB270507 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270507 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270507 Supplier Novus Biologicals Supplier No. NBP236549C
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTPN23 Polyclonal antibody specifically detects PTPN23 in Human samples. It is validated for Western Blot

Specifications

Antigen PTPN23
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.48, HDPTP, HD-PTPDKFZp564F0923, His domain-containing protein tyrosine phosphatase, His-domain protein tyrosine phosphatase, KIAA1471DKFZP564F0923, Protein tyrosine phosphatase TD14, protein tyrosine phosphatase, non-receptor type 23, PTP-TD14, tyrosine-protein phosphatase non-receptor type 23
Host Species Rabbit
Immunogen The immunogen recognized by the antibody maps to a region between residue 1586 to 1636 of human HisDomain Protein Tyrosine Phosphatase using the numbering given in entry NP_056281.1.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25930
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.