Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPN23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100047 Supplier Invitrogen™ Supplier No. PA5100047
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PTPN23.

Plays a role in sorting of endocytic ubiquitinated cargos into multivesicular bodies (MVBs) via its interaction with the ESCRT-I complex (endosomal sorting complex required for transport I), and possibly also other ESCRT complexes. May act as a negative regulator of Ras-mediated mitogenic activity. Plays a role in ciliogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN23
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PTPN23
Gene Accession No. O88902, Q6PB44, Q9H3S7
Gene Alias AI462446; HDPTP; HD-PTP; His domain-containing protein tyrosine phosphatase; Kiaa1471; Protein tyrosine phosphatase TD14; protein tyrosine phosphatase, non-receptor type 23; PTPN23; PTP-TD14; Tyrosine-protein phosphatase non-receptor type 23
Gene Symbols PTPN23
Host Species Rabbit
Immunogen A synthesized peptide derived from human PTPN23(Accession Q9H3S7), corresponding to amino acid residues P1553-H1603.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104831, 117552, 25930
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.