Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPN3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318412
This item is not returnable.
View return policy
Description
PTPN3 Polyclonal antibody specifically detects PTPN3 in Human samples. It is validated for Western BlotSpecifications
PTPN3 | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1:10000 - 1:25000 | |
DKFZp686N0569, EC 3.1.3.48, protein tyrosin phosphatase H1, protein tyrosine phosphatase, non-receptor type 3, Protein-tyrosine phosphatase H1, PTP-H1, PTPH1cytoskeletal-associated protein tyrosine phosphatase, tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 3 | |
Recognizes region between residues Between between 175 and 225.. | |
100 μg | |
Immunology, Signal Transduction | |
5774 | |
Store at 4°C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction