Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583414
Description
Immunogen sequence: TPNWRSPRLR HEIRKPRHSS ADNLANEMTY ITETEDVFYT YKGSLAPQDS DSEVSQNRSP HQESLSENNP AQSYLTQ.
Phosphorylation of receptors by protein kinases is a process that can be reversed by a group of enzymes called protein phosphatases. Coordinated control of kinases and phosphatases provides the cell with the capacity to rapidly switch between phosphorylated and dephosphorylated protein states in dynamic response to environmental stimuli. Activation of critical enzymes by kinase phosphorylation alone is not enough to provide adequate regulation - it is the combination with phosphatase dephosphorylation that effectively creates on/off switches to control cellular events. Errors in control, either through kinases or their counterpart phosphatases, can lead to unchecked cell growth attributable to human cancers and developmental disorders. Potential mechanisms to control dephosphorylation include changes in the expression of protein phosphatases, their subcellular localization, phosphorylation of phosphatase catalytic and regulatory subunits and regulation by endogenous phosphatase inhibitors. Most protein phosphatases are not stringently specific for their substrates. Consequently, changes in phosphatase activity may have a broad impact on dephosphorylation and turnover of phosphoproteins that are substrates for different kinases. This may be an important point of control to connect cellular circuitry of interrelated signaling pathways, and to synchronize physiological responses.
Specifications
PTPN3 | |
Polyclonal | |
Unconjugated | |
PTPN3 | |
9530011I20Rik; cytoskeletal-associated protein tyrosine phosphatase; protein tyrosine phosphatase, non-receptor type 3; protein-tyrosine phosphatase H1; PTPCL; PTPH1; PTP-H1; Ptpn3; Tyrosine-protein phosphatase non-receptor type 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5774 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P26045 | |
PTPN3 | |
Recombinant protein corresponding to Human PTPN3. Recombinant protein control fragment (Product #RP-98328). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction