Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550737 Supplier Invitrogen™ Supplier No. PA550737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PTPN4 protein.

The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This protein contains a C-terminal PTP domain and an N-terminal domain homologous to the band 4.1 superfamily of cytoskeletal-associated proteins. This PTP has been shown to interact with glutamate receptor delta 2 and epsilon subunits, and is thought to play a role in signaling downstream of the glutamate receptors through tyrosine dephosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PTPN4
Gene Accession No. P29074, Q9WU22
Gene Alias hPTP-MEG; MEG; megakaryocyte phosphatase; megakaryocyte protein-tyrosine phosphatase; protein tyrosine phosphatase MEG1; protein tyrosine phosphatase, non-receptor type 4; protein tyrosine phosphatase, non-receptor type 4 (megakaryocyte); protein-tyrosine phosphatase MEG1; Ptn4; PTP PTPMEG1; PTPase-MEG1; PTPMEG; PTPMEG1; Ptpn4; TEP; TEP/mPTPMEG; testis-enriched protein tyrosine phosphatase; Tyrosine-protein phosphatase non-receptor type 4
Gene Symbols PTPN4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the N terminal of human protein tyrosine phosphatase, non-receptor type 4 (megakaryocyte).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19258, 5775
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.