Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHP-1 Mouse anti-Human, Mouse, Rat, Clone: B5-A4, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP267086
Description
SHP-1 Monoclonal specifically detects SHP-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SHP-1 | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:200 | |
HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1 | |
Mouse | |
Protein A purified | |
Apoptosis, Phospho Specific, Protein Phosphatase | |
5777 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
B5-A4 | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide | |
PTPN6 | |
Recombinant protein within human SHP-1 aa 400-590/590. (SwissProt: P29350 Human) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction