Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPN7 Antibody (OTI3B10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273732F
Description
PTPN7 Monoclonal specifically detects PTPN7 in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PTPN7 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
PTPN7 | |
Full length recombinant protein of human PTPN7 (NP_542155) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG2b |
Immunocytochemistry, Immunofluorescence | |
OTI3B10 | |
Immunocytochemistry/Immunofluorescence | |
BPTP-4, FLJ18703, HEPTPnonreceptor-type, stress induced, LPTP, protein tyrosine phosphatase, non-receptor type 7, tyrosine-protein phosphatase non-receptor type 7 | |
Mouse | |
Affinity Purified | |
Cancer | |
5778.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction