Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPN7 Antibody (OTI3B10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP201340
Description
PTPN7 Monoclonal specifically detects PTPN7 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PTPN7 | |
Monoclonal | |
0.72 mg/mL | |
Western Blot 1:500-2000, Immunocytochemistry/Immunofluorescence 1:100 | |
PTPN7 | |
Full length human recombinant protein of human PTPN7(NP_542155) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG2b |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI3B10 | |
Unconjugated | |
BPTP-4, FLJ18703, HEPTPnonreceptor-type, stress induced, LPTP, protein tyrosine phosphatase, non-receptor type 7, tyrosine-protein phosphatase non-receptor type 7 | |
Mouse | |
44.8 kDa | |
0.1 mL | |
Cancer | |
5778 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction