Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN7 Monoclonal Antibody (OTI3B10), TrueMAB™, OriGene
SDP

Catalog No. 502540745
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0745 30 μL
50-167-4541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0745 Supplier OriGene Supplier No. TA503714S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTPN7 Monoclonal antibody specifically detects PTPN7 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PTPN7
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI3B10
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DAND5
Gene Accession No. Q8N907
Gene Alias BPTP-4; C920001D21Rik; dual specificity phosphatase 1; hematopoietic protein-tyrosine phosphatase; HEPTP; Lcptp; LC-PTP; LOW QUALITY PROTEIN: tyrosine-protein phosphatase non-receptor type 7; LPTP; protein tyrosine phosphatase non-receptor type 7; protein tyrosine phosphatase, non-receptor type 7; protein tyrosine phosphatase, non-receptor type 7 isoform 2; protein-tyrosine phosphatase LC-PTP; protein-tyrosine phosphatase, non-receptor type 7; protein-tyrosine phosphatase, nonreceptor-type, stress induced; Ptpn7; PTPNI; tyrosine-protein phosphatase non-receptor type 7
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 23-189 of human DAND5 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199699
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.