Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN7 Monoclonal Antibody (OTI3H8), TrueMAB™, OriGene
SDP

Catalog No. 502540777
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0777 30 μL
50-167-4502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0777 Supplier OriGene Supplier No. TA503662S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTPN7 Monoclonal antibody specifically detects PTPN7 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PTPN7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3H8
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TMED1
Gene Accession No. Q13445
Gene Alias BPTP-4; C920001D21Rik; dual specificity phosphatase 1; hematopoietic protein-tyrosine phosphatase; HEPTP; Lcptp; LC-PTP; LOW QUALITY PROTEIN: tyrosine-protein phosphatase non-receptor type 7; LPTP; protein tyrosine phosphatase non-receptor type 7; protein tyrosine phosphatase, non-receptor type 7; protein tyrosine phosphatase, non-receptor type 7 isoform 2; protein-tyrosine phosphatase LC-PTP; protein-tyrosine phosphatase, non-receptor type 7; protein-tyrosine phosphatase, nonreceptor-type, stress induced; Ptpn7; PTPNI; tyrosine-protein phosphatase non-receptor type 7
Host Species Mouse
Immunogen Full length human recombinant protein of human TMED1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11018
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.