Learn More
Description
Specifications
Specifications
| Antigen | PTPRB |
| Applications | Flow Cytometry, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:16000 |
| Gene Alias | DKFZp686E2262, DKFZp686H15164, EC 3.1.3.48, FLJ44133, HPTPB, HPTP-BETA, MGC142023, MGC59935, protein tyrosine phosphatase, receptor type, B, protein tyrosine phosphatase, receptor type, beta polypeptide, Protein-tyrosine phosphatase beta, PTPB, receptor-type tyrosine-protein phosphatase beta, R-PTP-BETA, Vascular endothelial protein tyrosine phosphatase, VEPTP, VE-PTP |
| Host Species | Goat |
| Immunogen | Peptide with sequence C-ENHSFSQERTVPDK corresponding to internal region according to NP_001103224.1, NP_002828.3. |
| Purification Method | Immunogen affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
