Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRCAP Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB272884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272884 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272884 Supplier Novus Biologicals Supplier No. NBP236565AF532
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTPRCAP Polyclonal antibody specifically detects PTPRCAP in Human samples. It is validated for Western Blot

Specifications

Antigen PTPRCAP
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CD45-APCD45 associated protein, CD45-associated protein, LPAPMGC138603, Lymphocyte phosphatase-associated phosphoprotein, MGC138602, protein tyrosine phosphatase receptor type C-associated protein, protein tyrosine phosphatase, receptor type, c polypeptide-associated protein, protein tyrosine phosphatase, receptor type, C-associated protein, PTPRC-associated protein
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 156 to 206 of human Lymphocyte Phosphatase-Associated Phosphoprotein using the numbering given in entry NP_005599.1.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5790
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.