Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRCAP Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5118224
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118224 Supplier Thermo Scientific Supplier No. PA5118224
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MSK-1 is a mitogen and stress activated protein kinase-1 which belongs to the AGC family of kinases and is related in structure to the ribosomal p70 S6 kinase subfamily. MSK-1 can be activated by ERK1/2 and SAPK2 /p38 MAP kinase. It is also known to be required for the phosphorylation of CREB, ATF1 H3 and HMG-14 in response to mitogen and stress. Similar to RSK, MSK-1 contains two kinase domains (N-term and a C-term). Once phosphorylated on Thr581 and Ser360 by ERK1/2 and SAPK2/p38, MSK-1 autophosphorylate on at least 5 sites. Of these autophosphorylation sites Ser212 and Ser376 get phosphorylated by the C-terminal kinase domain of MSK-1 which is essential for the catalytic activity of the N-terminal kinase domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRCAP
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene PTPRCAP
Gene Accession No. Q14761
Gene Alias AI605264; CD45 associated protein; CD-45-AP; CD45-AP; CD45-associated protein; LPAP; LSM-1; Lymphocyte phosphatase-associated phosphoprotein; protein tyrosine phosphatase receptor type C-associated protein; protein tyrosine phosphatase, receptor type C associated protein; protein tyrosine phosphatase, receptor type, C polypeptide-associated protein; protein tyrosine phosphatase, receptor type, C-associated protein; Ptprcap; PTPRC-associated protein
Gene Symbols PTPRCAP
Host Species Rabbit
Immunogen E. coli-derived Human PTPRCAP fragment
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5790
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.