Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597116 Supplier Invitrogen™ Supplier No. PA597116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QITTIGNLVP QKTYSVKVLA FTSIGDGPLS SDIQVITQTG VPGQPLNFKA EPESETSILL SWTPPRSDTI ANYELVYKDG EHGEEQRITI EPGTSYRLQG; Positive Samples: HT-1080, DU145, B cells, Mouse heart; Cellular Location: Membrane, Single-pass type I membrane protein.

PTPRD is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular region, a single transmembrane segment and two tandem intracytoplasmic catalytic domains, and thus represents a receptor-type PTP. The extracellular region of this protein is composed of three Ig-like and eight fibronectin type III-like domains. Studies of the similar genes in chicken and fly suggest the role of this PTP is in promoting neurite growth, and regulating neurons axon guidance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRD
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PTPRD
Gene Accession No. P23468, Q64487
Gene Alias 1110002J03Rik; 3000002J10Rik; B230219D21Rik; HPTP; HPTPD; HPTPDELTA; protein tyrosine phosphatase, receptor type D; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, delta polypeptide; Protein-tyrosine phosphatase delta; PTPD; Ptprd; receptor-type tyrosine-protein phosphatase delta; RPTPDELTA; R-PTP-delta
Gene Symbols PTPRD
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19266, 5789
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.