Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPRD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA526593
Description
PTPRD Polyclonal Antibody for Western Blot
PTPRD is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular region, a single transmembrane segment and two tandem intracytoplasmic catalytic domains, and thus represents a receptor-type PTP. The extracellular region of this protein is composed of three Ig-like and eight fibronectin type III-like domains. Studies of the similar genes in chicken and fly suggest the role of this PTP is in promoting neurite growth, and regulating neurons axon guidance.
Specifications
| PTPRD | |
| Polyclonal | |
| Unconjugated | |
| PTPRD | |
| 1110002J03Rik; 3000002J10Rik; B230219D21Rik; HPTP; HPTPD; HPTPDELTA; protein tyrosine phosphatase, receptor type D; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, delta polypeptide; Protein-tyrosine phosphatase delta; PTPD; Ptprd; receptor-type tyrosine-protein phosphatase delta; RPTPDELTA; R-PTP-delta | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5789 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P23468 | |
| PTPRD | |
| KLH conjugated synthetic peptide between 343-371 amino acids from the N-terminal region of human PTPRD. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction