Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRE Monoclonal Antibody (OTI3H3), TrueMAB™, OriGene
SDP

Catalog No. 502542440
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2440 30 μL
50-167-2375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2440 Supplier OriGene Supplier No. TA501234S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTPRE Monoclonal antibody specifically detects PTPRE in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen PTPRE
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI3H3
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PTPRE
Gene Accession No. P23469
Gene Alias HPTPE; protein tyrosine phosphatase epsilon; protein tyrosine phosphatase epsilon-like 1; protein tyrosine phosphatase epsilon-like 2; protein tyrosine phosphatase, receptor type E; protein tyrosine phosphatase, receptor type, E; protein tyrosine phosphatase, receptor type, epsilon; protein tyrosine phosphatase, receptor type, epsilon polypeptide; protein-tyrosine phosphatase epsilon; PTP epsilon; PTPE; PTPepsilon; PTPRE; PTPvarepsilon; receptor-type tyrosine-protein phosphatase epsilon; RPTPepsilon; R-PTP-epsilon
Host Species Mouse
Immunogen Full length human recombinant protein of human PTPRE produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5791
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.