Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587304 Supplier Invitrogen™ Supplier No. PA587304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. Two alternatively spliced transcript variants of this gene have been reported, one of which encodes a receptor-type PTP that possesses a short extracellular domain, a single transmembrane region, and two tandem intracytoplasmic catalytic domains; Another one encodes a PTP that contains a distinct hydrophilic N-terminus, and thus represents a nonreceptor-type isoform of this PTP. Studies of the similar gene in mice suggested the regulatory roles of this PTP in RAS related signal transduction pathways, cytokines induced SATA signaling, as well as the activation of voltage-gated K+ channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPRE
Gene Accession No. B2GV87, P23469, P49446
Gene Alias HPTPE; protein tyrosine phosphatase epsilon; protein tyrosine phosphatase epsilon-like 1; protein tyrosine phosphatase epsilon-like 2; protein tyrosine phosphatase, receptor type E; protein tyrosine phosphatase, receptor type, E; protein tyrosine phosphatase, receptor type, epsilon; protein tyrosine phosphatase, receptor type, epsilon polypeptide; protein-tyrosine phosphatase epsilon; PTP epsilon; PTPE; PTPepsilon; PTPRE; PTPvarepsilon; receptor-type tyrosine-protein phosphatase epsilon; RPTPepsilon; R-PTP-epsilon
Gene Symbols PTPRE
Host Species Rabbit
Immunogen Recombinant full length Human PTPRE.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114767, 19267, 5791
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.