Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRF Monoclonal Antibody (6D7C7)
GREENER_CHOICE

Catalog No. PIMA538615
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538615 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538615 Supplier Invitrogen™ Supplier No. MA538615
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

Possible cell adhesion receptor. It possesses an intrinsic protein tyrosine phosphatase activity (PTPase).The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRF
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6D7C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PTPRF
Gene Accession No. P10586
Gene Alias AA591035; BNAH2; FLJ43335; FLJ45062; FLJ45567; LAR; LAR protein; LARFN5C; LARS; LCA homolog; LCA-homolog; leukocyte antigen-related (LAR) PTP receptor; leukocyte antigen-related tyrosine phosphatase; leukocyte common antigen related; protein tyrosine phosphatase receptor-type F; protein tyrosine phosphatase, receptor type F; protein tyrosine phosphatase, receptor type, F; protein tyrosine phosphatase, receptor type, F polypeptide; Ptprf; PTPRF protein; receptor-linked protein-tyrosine phosphatase LAR; receptor-type tyrosine-protein phosphatase F; RPTP-LAR
Gene Symbols PTPRF
Host Species Mouse
Immunogen Purified recombinant fragment of human PTPRF (aa: extra 1104-1163) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5792
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.