Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRF Monoclonal Antibody (N165/38), APC
GREENER_CHOICE

Catalog No. PIMA545637
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545637 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545637 Supplier Invitrogen™ Supplier No. MA545637
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

97% identical in both rat and mouse. >80% identity with PTPRD and PTPRS. >50% identity with PTPRM and PTPRK. 1 μg/mL of MA5-45637 was sufficient for detection of LAR/PTPRF in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 85kDa (full length protein is 210 kDa - smaller due to proteolysis into P-subunit containing transmembrane and intracellular domains. This antibody was formerly sold as clone S165-38.

Possible cell adhesion receptor. It possesses an intrinsic protein tyrosine phosphatase activity (PTPase).The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRF
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N165/38
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene PTPRF
Gene Accession No. A2A8L5, P10586, Q64604
Gene Alias AA591035; BNAH2; FLJ43335; FLJ45062; FLJ45567; LAR; LAR protein; LARFN5C; LARS; LCA homolog; LCA-homolog; leukocyte antigen-related (LAR) PTP receptor; leukocyte antigen-related tyrosine phosphatase; leukocyte common antigen related; protein tyrosine phosphatase receptor-type F; protein tyrosine phosphatase, receptor type F; protein tyrosine phosphatase, receptor type, F; protein tyrosine phosphatase, receptor type, F polypeptide; Ptprf; PTPRF protein; receptor-linked protein-tyrosine phosphatase LAR; receptor-type tyrosine-protein phosphatase F; RPTP-LAR
Gene Symbols PTPRF
Host Species Mouse
Immunogen Fusion protein amino acids 1315-1607 (cytoplasmic C-terminus) of human LAR.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19268, 360406, 5792
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.