Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPRH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP325081
Description
PTPRH Polyclonal antibody specifically detects PTPRH in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
PTPRH | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
EC 3.1.3.48, FLJ39938, MGC133058, MGC133059, protein tyrosine phosphatase, receptor type, H, receptor-type tyrosine-protein phosphatase H, R-PTP-H, SAP1, SAP-1, Stomach cancer-associated protein tyrosine phosphatase 1, Transmembrane-type protein-tyrosine phosphatase type H | |
This antibody has been engineered to specifically recognize the recombinant protein PTPRH using the following amino acid sequence: QANWVNQTSRTNETWYKVEALEPGTLYNFTVWAERNDVASSTQSLCASTYPDTVTITSCVSTSAGYGVNLIWSCPQGGYEAFELEVGGQRGSQDRSSCGEAVSVLGLGPARSYPATITTIWDGMKVVSHSVVC | |
100 μL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
5794 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction