Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPRH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144686
Description
Immunogen sequence: QANWVNQTSR TNETWYKVEA LEPGTLYNFT VWAERNDVAS STQSLCASTY PDTVTITSCV STSAGYGVNL IWSCPQGGYE AFELEVGGQR GSQDRSSCGE AVSVLGLGPA RSYPATITTI WDGMKVVSHS VVC Antigen sequence identity: Mouse 46%; Rat 45%.
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracytoplasmic catalytic domain, and thus represents a receptor-type PTP. The extracellular region contains eight fibronectin type III-like repeats and multiple N-glycosylation sites. The gene was shown to be expressed primarily in brain and liver, and at a lower level in heart and stomach. It was also found to be expressed in several cancer cell lines, but not in the corresponding normal tissues.
Specifications
PTPRH | |
Polyclonal | |
Unconjugated | |
PTPRH | |
Bem2; brain-enriched membrane-associated protein tyrosine BEM-2; protein tyrosine phosphatase, receptor type H; protein tyrosine phosphatase, receptor type, H; PTPRH; receptor-type tyrosine-protein phosphatase H; R-PTP-H; SAP1; SAP-1; Stomach cancer-associated protein tyrosine phosphatase 1; transmembrane-type protein-tyrosine phosphatase type H | |
Rabbit | |
Affinity chromatography | |
RUO | |
5794 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HD43 | |
PTPRH | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 615-747. Recombinant protein control fragment (Product #RP-109971). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction