Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRH Polyclonal Antibody

Catalog No. PIPA5121662
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121662 20 μL
PIPA5119375 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121662 Supplier Invitrogen™ Supplier No. PA5121662
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTPRH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracytoplasmic catalytic domain, and thus represents a receptor-type PTP. The extracellular region contains eight fibronectin type III-like repeats and multiple N-glycosylation sites. The gene was shown to be expressed primarily in brain and liver, and at a lower level in heart and stomach. It was also found to be expressed in several cancer cell lines, but not in the corresponding normal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PTPRH
Gene Accession No. E9Q0N2
Gene Alias Bem2; brain-enriched membrane-associated protein tyrosine BEM-2; protein tyrosine phosphatase, receptor type H; protein tyrosine phosphatase, receptor type, H; PTPRH; receptor-type tyrosine-protein phosphatase H; R-PTP-H; SAP1; SAP-1; Stomach cancer-associated protein tyrosine phosphatase 1; transmembrane-type protein-tyrosine phosphatase type H
Gene Symbols PTPRH
Host Species Rabbit
Immunogen Recombinant protein Tyrosine Phosphatase Receptor Type H. The antigen corresponds to amino acid range 685-909 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 545902
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.