Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563026 Supplier Invitrogen™ Supplier No. PA563026
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTHMVNAMDR SYADQSTLHA EDPLSITFMD QHNFSPRYEN HSATAESSRL LDVPRYLCE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Protein tyrosine phosphatases, or PTPs, are type I transmembrane proteins, membrane associated proteins or proteins localized in nuclei. Examples of transmembrane PTPs are LAR, PTP alpha, PTP beta, PTP gamma, PTP delta, PTP epsilon, PTP zeta, PTPk and PTP-mu. Transmembrane PTPs play diverse roles during development and in adult tissues. Immunodepletion studies have suggested LAR to be a regulator of Insulin receptor phosphorylation. PTP alpha activity is increased twofold in response to phorbol ester stimulation, resulting in serine phosphorylation either directly or indirectly by members of the PKC family. Overexpression of v-H-ras and Neu, but not Myc or Int2, in mammary tumors has been shown to induce PTP epsilon expression. An alternative splicing event leads to a nervous tissue-specific chondroitin sulfate proteoglycan called phosphacan, which represents the amino terminal portion of PTP zeta. PTPk and PTP-mu share a conserved amino terminal 160 amino acid MAM domain which facilitates homophilic binding. PTP-mu localizes to points of cell contact and may be involved in regulating the assembly and disassembly of cadherin/catenin complexes in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRK
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPRK
Gene Accession No. Q15262
Gene Alias AI853699; dJ480J14.2.1 (protein tyrosine phosphatase, receptor type, K (R-PTP-KAPPA, protein tyrosine phosphatase kappa , protein tyrosine phosphatase kappa; protein tyrosine phosphatase, receptor type K; protein tyrosine phosphatase, receptor type, K; protein tyrosine phosphatase, receptor type, K, extracellular region; protein-tyrosine phosphatase kappa; protein-tyrosine phosphatase, receptor type, kappa; Ptpk; PTPRK; receptor-like protein tyrosine phosphatase kappa extracellular region (RPTPK); receptor-type tyrosine-protein phosphatase kappa; Rptpk; R-PTPk; R-PTP-k; RPTPkappa; R-PTP-kappa
Gene Symbols PTPRK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTPRK. Recombinant protein control fragment (Product #RP-101384).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5796
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.