Learn More
PTPRN2 Monoclonal Antibody (8E12)

Description
PTPRN2 Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | PTPRN2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 8E12 |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PTPRN2 |
| Gene Accession No. | Q92932 |
| Gene Alias | 4930425H11Rik; EC 3.1.3.48; IA-2 beta; IA2beta; IA-2beta; IA-2beta60; IA-2beta64; IA-2beta71; IAR; IAR/receptor-like protein-tyrosine phosphatase; ICAAR; Islet cell autoantigen-related protein; KIAA0387; mKIAA0387; phogrin; Phol; protein tyrosine phosphatase receptor pi; protein tyrosine phosphatase receptor-type N polypeptide 2; protein tyrosine phosphatase, receptor type N2; protein tyrosine phosphatase, receptor type, N polypeptide 2; protein tyrosine phosphatase, receptor-type, N polypeptide 2; protein tyrosine phosphatase-NP; PTP IA-2beta; PTP NE-6; PTPNE6; PTP-NP; PTPRN2; PTPRP; Receptor-type tyrosine-protein phosphatase N2; R-PTP-N2; tyrosine phosphatase IA-2 beta |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.