Learn More
Invitrogen™ PTPRN2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total PTPRN2 protein.
Specifications
Specifications
| Antigen | PTPRN2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PTPRN2 |
| Gene Accession No. | P80560, Q92932 |
| Gene Alias | 4930425H11Rik; EC 3.1.3.48; IA-2 beta; IA2beta; IA-2beta; IA-2beta60; IA-2beta64; IA-2beta71; IAR; IAR/receptor-like protein-tyrosine phosphatase; ICAAR; Islet cell autoantigen-related protein; KIAA0387; mKIAA0387; phogrin; Phol; protein tyrosine phosphatase receptor pi; protein tyrosine phosphatase receptor-type N polypeptide 2; protein tyrosine phosphatase, receptor type N2; protein tyrosine phosphatase, receptor type, N polypeptide 2; protein tyrosine phosphatase, receptor-type, N polypeptide 2; protein tyrosine phosphatase-NP; PTP IA-2beta; PTP NE-6; PTPNE6; PTP-NP; PTPRN2; PTPRP; Receptor-type tyrosine-protein phosphatase N2; R-PTP-N2; tyrosine phosphatase IA-2 beta |
| Gene Symbols | PTPRN2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.