Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590035 Supplier Invitrogen™ Supplier No. PA590035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAPSSVPRGR QLPGRLGCLL EEGLCGASEA CVNDGVFGRC QKVPAMDFYR YEVSPVALQR LRVALQKLSG TGFTWQDDYT QYVMDQELAD LPKTYLRRPE ASSPARPSKH SVGSERRYSR EGGAALANAL RRHLPFLEAL SQAPASDVLA RTHTAQDRPP AEGDDRFSES ILTYVAHTSA LTYPPGSRTQ LREDLLPRTL GQLQPDELSP KVDSGVDRHH L; Positive Samples: SH-SY5Y, SGC-7901, Mouse pancreas, Rat pancreas; Cellular Location: Cytoplasmic vesicle, Single-pass type I membrane protein, secretory vesicle, secretory vesicle membrane, synaptic vesicle membrane.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domain, and thus represents a receptor-type PTP. The catalytic domain of this PTP is most closely related to PTPRN/IA-2beta. This PTP and PTPRN are both found to be major autoantigens associated with insulin-dependent diabetes mellitus. Three alternatively spliced transcript variants of this gene, which encode distinct proteins, have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRN2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene PTPRN2
Gene Accession No. P80560, Q63475, Q92932
Gene Alias 4930425H11Rik; EC 3.1.3.48; IA-2 beta; IA2beta; IA-2beta; IA-2beta60; IA-2beta64; IA-2beta71; IAR; IAR/receptor-like protein-tyrosine phosphatase; ICAAR; Islet cell autoantigen-related protein; KIAA0387; mKIAA0387; phogrin; Phol; protein tyrosine phosphatase receptor pi; protein tyrosine phosphatase receptor-type N polypeptide 2; protein tyrosine phosphatase, receptor type N2; protein tyrosine phosphatase, receptor type, N polypeptide 2; protein tyrosine phosphatase, receptor-type, N polypeptide 2; protein tyrosine phosphatase-NP; PTP IA-2beta; PTP NE-6; PTPNE6; PTP-NP; PTPRN2; PTPRP; Receptor-type tyrosine-protein phosphatase N2; R-PTP-N2; tyrosine phosphatase IA-2 beta
Gene Symbols PTPRN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-242 of human PTPRN2 (NP_5708582).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19276, 29714, 5799
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.