Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599696 Supplier Invitrogen™ Supplier No. PA599696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PTP IA-2beta.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domain, and thus represents a receptor-type PTP. The catalytic domain of this PTP is most closely related to PTPRN/IA-2beta. This PTP and PTPRN are both found to be major autoantigens associated with insulin-dependent diabetes mellitus. Three alternatively spliced transcript variants of this gene, which encode distinct proteins, have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRN2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PTPRN2
Gene Accession No. P80560, Q63475, Q92932
Gene Alias 4930425H11Rik; EC 3.1.3.48; IA-2 beta; IA2beta; IA-2beta; IA-2beta60; IA-2beta64; IA-2beta71; IAR; IAR/receptor-like protein-tyrosine phosphatase; ICAAR; Islet cell autoantigen-related protein; KIAA0387; mKIAA0387; phogrin; Phol; protein tyrosine phosphatase receptor pi; protein tyrosine phosphatase receptor-type N polypeptide 2; protein tyrosine phosphatase, receptor type N2; protein tyrosine phosphatase, receptor type, N polypeptide 2; protein tyrosine phosphatase, receptor-type, N polypeptide 2; protein tyrosine phosphatase-NP; PTP IA-2beta; PTP NE-6; PTPNE6; PTP-NP; PTPRN2; PTPRP; Receptor-type tyrosine-protein phosphatase N2; R-PTP-N2; tyrosine phosphatase IA-2 beta
Gene Symbols PTPRN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PTPRN2(Accession Q92932), corresponding to amino acid residues P205-P255.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19276, 29714, 5799
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.