Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556964
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556964 Supplier Invitrogen™ Supplier No. PA556964
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TRVNISYWEG KDFRTMLYKD FFKGKTVFNH WLPGMCYSNI TFQLVSEATF NKSTLVEYSG VSHEPKQHRT APYPPQNISV RIVNLNKNNW EEQSGNFPEE SFMRSQDTIG KEKLFHFTEE TPEIPSGNIS SGWP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.

Glomerular epithelial protein 1 (GLEPP1), also designated protein tyrosine phosphatase U2 (PTPase U2) or receptor-type tyrosine-protein phosphatase O (PTPRO), belongs to the protein-tyrosine phosphatase family. GLEPP1 is a Type I membrane protein containing 8 fibronectin type-III domains and 1 tyrosine-protein phosphatase domain. GLEPP1 is expressed primarily in the glomeruli of the kidney, but is also detected in placenta, lung and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRO
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPRO
Gene Accession No. Q16827
Gene Alias D28; FMI; GLEPP1; Glomerular epithelial protein 1; hPTP-J; NPHS6; osteoclastic transmembrane protein-tyrosine phosphatase; PCP2; PCP-2; phosphotyrosine phosphatase U2; protein tyrosine phosphatase PTP-U2; protein tyrosine phosphatase U2; protein tyrosine phosphatase, non-receptor type 15; protein tyrosine phosphatase, receptor type O; protein tyrosine phosphatase, receptor type, O; PTP phi; PTP pi; PTPase U2; PTPBK; PTP-J; Ptpn15; PTPoc; PTP-OC; PTPphi; Ptpro; PTP-RO; PTPROT; PTPRU; PTPU2; PTP-U2; receptor protein tyrosine phosphatase RO; receptor-type protein tyrosine phosphatase D30; receptor-type tyrosine-protein phosphatase O; RPTP-BK; R-PTP-O; R-PTP-psi; R-PTP-U
Gene Symbols PTPRO
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTPRO. Recombinant protein control fragment (Product #RP-101062).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5800
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.