Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547487
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547487 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547487 Supplier Invitrogen™ Supplier No. PA547487
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody detects the endogenous cleaved extracellular domain of human PTP sigma /PTPRS in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human LAR, PTPR beta, PTPR gamma, PTPR mu, or PTPR kappa is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

PTPRS cell surface receptor that binds to glycosaminoglycans, including chondroitin sulfate proteoglycans and heparan sulfate proteoglycan. Binding to chondroitin sulfate and heparan sulfate proteoglycans has opposite effects on PTPRS oligomerization and regulation of neurite outgrowth. Contributes to the inhibition of neurite and axonal outgrowth by chondroitin sulfate proteoglycans, also after nerve transection. Plays a role in stimulating neurite outgrowth in response to the heparan sulfate proteoglycan GPC2. Required for normal brain development, especially for normal development of the pituitary gland and the olfactory bulb. Functions as tyrosine phosphatase. Mediates dephosphorylation of NTRK1, NTRK2 and NTRK3. Plays a role in down-regulation of signaling cascades that lead to the activation of Akt and MAP kinases. Down-regulates TLR9-mediated activation of NF-kappa-B, as well as production of TNF, interferon alpha and interferon beta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRS
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Ptprs
Gene Accession No. Q13332
Gene Alias AL022616; Larptp2; LAR-PTP2; Leukocyte common antigen-related protein-tyrosine phosphatase 2; leukocyte common antigen-related proten-tyrosine phosphatase 2; protein tyrosine phosphatase PTPsigma; protein tyrosine phosphatase, receptor type S; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, S; PTP; PTP-NU3; PTPNU-3; Ptprd; Ptprs; PTPsigma; Ptpsigma.; Ptpt9; Receptor-type tyrosine-protein phosphatase S; Receptor-type tyrosine-protein phosphatase sigma; Rptpd; R-PTP-S; RPTPsigma; R-PTP-sigma; tissue-specific protein tyrosine phosphatase
Gene Symbols Ptprs
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human PTP sigma/PTPRS Glu30-Gly1260.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5802
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.