Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597164
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597164 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597164 Supplier Invitrogen™ Supplier No. PA597164
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNRGSSLGGL QQTVTAWTAF NLLNGKPSVA PKPDADGFIM VYLPDGQSPV PVQSYFIVMV PLRKSRGGQF LTPLGSPEDM DLEELIQDIS RLQRRSLRHS RQLEVPRPYI AARFSVLPPT FHPGDQKQYG GFDNRGLEPG HRYVLFVLAV LQKSEPTFAA SPFSDPFQLD NPDPQPIVDG E; Positive Samples: Rat testis; Cellular Location: Membrane, Single-pass type I membrane protein.

PTPRS cell surface receptor that binds to glycosaminoglycans, including chondroitin sulfate proteoglycans and heparan sulfate proteoglycan. Binding to chondroitin sulfate and heparan sulfate proteoglycans has opposite effects on PTPRS oligomerization and regulation of neurite outgrowth. Contributes to the inhibition of neurite and axonal outgrowth by chondroitin sulfate proteoglycans, also after nerve transection. Plays a role in stimulating neurite outgrowth in response to the heparan sulfate proteoglycan GPC2. Required for normal brain development, especially for normal development of the pituitary gland and the olfactory bulb. Functions as tyrosine phosphatase. Mediates dephosphorylation of NTRK1, NTRK2 and NTRK3. Plays a role in down-regulation of signaling cascades that lead to the activation of Akt and MAP kinases. Down-regulates TLR9-mediated activation of NF-kappa-B, as well as production of TNF, interferon alpha and interferon beta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRS
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ptprs
Gene Accession No. Q64605
Gene Alias AL022616; Larptp2; LAR-PTP2; Leukocyte common antigen-related protein-tyrosine phosphatase 2; leukocyte common antigen-related proten-tyrosine phosphatase 2; protein tyrosine phosphatase PTPsigma; protein tyrosine phosphatase, receptor type S; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, S; PTP; PTP-NU3; PTPNU-3; Ptprd; Ptprs; PTPsigma; Ptpsigma.; Ptpt9; Receptor-type tyrosine-protein phosphatase S; Receptor-type tyrosine-protein phosphatase sigma; Rptpd; R-PTP-S; RPTPsigma; R-PTP-sigma; tissue-specific protein tyrosine phosphatase
Gene Symbols Ptprs
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1100-1280 of human PTPRS (NP_0028413).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25529
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.