Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRT Polyclonal Antibody, Invitrogen™

Catalog No. PIPA518304
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518304 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518304 Supplier Thermo Scientific Supplier No. PA518304

Goat Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

PTPRT Polyclonal antibody specifically detects PTPRT in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PTPRT
Gene Accession No. O14522
Gene Alias KIAA0283; mKIAA0283; mRPTPrho; protein tyrosine phosphatase, receptor type T; protein tyrosine phosphatase, receptor type, T; PTPRT; receptor protein tyrosine phosphatase; receptor protein tyrosine phosphatase-rho; receptor-type tyrosine-protein phosphatase rho; Receptor-type tyrosine-protein phosphatase T; RP5-1121H13.2; RPTPmam4; RPTP-rfo; RPTPrho; RPTP-rho; R-PTP-T
Gene Symbols PTPRT
Host Species Goat
Immunogen Synthetic peptide sequence (CNTWEKPMLDQAV) corresponding to the internal amino acids of PTPRT
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11122
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.