Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRU Monoclonal Antibody (1A6E5)
GREENER_CHOICE

Catalog No. PIMA531875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531875 Supplier Invitrogen™ Supplier No. MA531875
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31875 has been tested in indirect ELISA.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and two tandem intracellular catalytic domains, and thus represents a receptor-type PTP. The extracellular region contains a meprin-A5 antigen-PTP (MAM) domain, Ig-like and fibronectin type III-like repeats. This PTP was thought to play roles in cell-cell recognition and adhesion. Studies of the similar gene in mice suggested the role of this PTP in early neural development. The expression of this gene was reported to be regulated by phorbol myristate acetate (PMA) or calcium ionophore in Jurkat T lymphoma cells. Alternatively spliced transcript variants have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRU
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A6E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PTPRU
Gene Accession No. Q92729
Gene Alias FMI; Ftp-1; GLEPP1; hPTP-J; Pancreatic carcinoma phosphatase 2; Pcp2; PCP-2; pi R-PTP-Psi; protein tyrosine phosphatase, receptor type U; protein tyrosine phosphatase, receptor type, L; protein tyrosine phosphatase, receptor type, U; protein-tyrosine phosphatase J; Protein-tyrosine phosphatase pi; protein-tyrosine phosphatase receptor omicron; Protein-tyrosine phosphatase-lamda; PTP; PTP pi; Ptpf; PTP-J; PTPlambda; PTP-lambda; PTP-PI; PTPPSI; Ptprl; PTPRO; PTP-RO; PTPRU; PTPU2; Receptor protein tyrosine phosphatase hPTP-J; receptor-type protein-tyrosine phosphatase psi; Receptor-type tyrosine-protein phosphatase U; RPTPlambda; R-PTP-psi; R-PTP-U
Gene Symbols PTPRU
Host Species Mouse
Immunogen Purified recombinant fragment of human PTPRU (AA: extra 579-749) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.