Learn More
Invitrogen™ PTPRU Monoclonal Antibody (764209)

Description
This antibody detects human and rat PTPRU in Western blots and recombinant human PTPRU in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) PTPRK, rhPTPRM, or rhPTPRT is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | PTPRU |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 764209 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | PTPRU |
| Gene Accession No. | Q92729 |
| Gene Alias | FMI; Ftp-1; GLEPP1; hPTP-J; Pancreatic carcinoma phosphatase 2; Pcp2; PCP-2; pi R-PTP-Psi; protein tyrosine phosphatase, receptor type U; protein tyrosine phosphatase, receptor type, L; protein tyrosine phosphatase, receptor type, U; protein-tyrosine phosphatase J; Protein-tyrosine phosphatase pi; protein-tyrosine phosphatase receptor omicron; Protein-tyrosine phosphatase-lamda; PTP; PTP pi; Ptpf; PTP-J; PTPlambda; PTP-lambda; PTP-PI; PTPPSI; Ptprl; PTPRO; PTP-RO; PTPRU; PTPU2; Receptor protein tyrosine phosphatase hPTP-J; receptor-type protein-tyrosine phosphatase psi; Receptor-type tyrosine-protein phosphatase U; RPTPlambda; R-PTP-psi; R-PTP-U |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.