Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101832
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101832 Supplier Invitrogen™ Supplier No. PA5101832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PTPRZ1.

Protein tyrosine phosphatase that negatively regulates oligodendrocyte precursor proliferation in the embryonic spinal cord. Required for normal differentiation of the precursor cells into mature, fully myelinating oligodendrocytes. May play a role in protecting oligondendrocytes against apoptosis. May play a role in the establishment of contextual memory, probably via the dephosphorylation of proteins that are part of important signaling cascades.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRZ
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PTPRZ1
Gene Accession No. B9EKR1, P23471, Q62656
Gene Alias 3F8 chondroitin sulfate proteoglycan; 3H1 keratan sulfate proteoglycan; 6B4 proteoglycan; AI850339; DSD-1-PG; HPTPZ; HPTPzeta; HTPZP2; phosphacan; protein tyrosine phosphatase, receptor type Z, polypeptide 1; protein tyrosine phosphatase, receptor type Z1; protein tyrosine phosphatase, receptor-type, Z polypeptide 1; protein tyrosine phosphatase, receptor-type, zeta polypeptide 1; protein tyrosine phosphate, receptor type, Z; Protein-tyrosine phosphatase receptor type Z polypeptide 1; Protein-tyrosine phosphatase receptor type Z polypeptide 2; PTP zeta; PTP18; PTPbeta; PTPRZ; PTPRZ1; PTPRZ2; Ptpz; PTPzeta; PTP-ZETA; PTPzeta-A; PTPzeta-B; PTPzeta-S; receptor-type tyrosine phosphatase beta/zeta; receptor-type tyrosine-protein phosphatase zeta; RPTPB; RPTPbeta; RPTPz; R-PTP-zeta; R-PTP-zeta-2
Gene Symbols PTPRZ1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PTPRZ1(Accession P23471), corresponding to amino acid residues F121-K171.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19283, 25613, 5803
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.