Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTRF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PTRF |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PTRF Polyclonal specifically detects PTRF in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PTRF | |
Polyclonal | |
Rabbit | |
Diabetes Research, Lipid and Metabolism | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
284119 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VSVNVKTVRGSLERQAGQIKKLEVNEAELLRRRNFKVMIYQDEVKLPAKLSISKSLKESEALPEKEGEELGEGERPEEDAAALELSSD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
CAVIN, Cavin-1, CGL4, FLJ90031, polymerase I and transcript release factor, RNA polymerase I and transcript release factor, TTF-I interacting peptide 12 | |
PTRF | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title