Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTRF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583981
Description
Immunogen sequence: EPSSAGAQAA EEPSGAGSEE LIKSDQVNGV LVLSLLDKII GAVDQIQLTQ AQLEERQAEM EGAVQSIQGE LSKLGK.
The polymerase I and transcript release factor (PTRF) enables the dissociation of paused ternary polymerase I transcription complexes from the 3' end of pre-rRNA transcripts, regulating rRNA transcription by promoting the dissociation of transcription complexes and the re-initiation of polymerase I on nascent rRNA transcripts. PTRF also localizes to caveolae at the plasma membrane and is thought to play a critical role in the formation of caveolae and the stabilization of caveolins, translocating from caveolae to the cytoplasm after insulin stimulation. PTRF is also thought to modify lipid metabolism and insulin-regulated gene expression. Mutations in this gene result in a disorder characterized by generalized lipodystrophy and muscular dystrophy.
Specifications
PTRF | |
Polyclonal | |
Unconjugated | |
CAVIN1 | |
2310075E07Rik; AW546441; caveolae associated protein 1; caveolae-associated protein 1; CAVIN; CAVIN1; cavin-1; Cav-p60; CGL4; FKSG13; FLJ90031; Polymerase I and transcript release factor; PTRF; ptrf {ECO:0000312; RGD:1307685}; RNA polymerase I and transcript release factor; TTF-I interacting peptide 12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284119 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NZI2 | |
CAVIN1 | |
Recombinant protein corresponding to Human CAVIN1. Recombinant protein control fragment (Product #RP-99929). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction