Learn More
PTRF Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301270AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Termination of transcription by RNA polymerase I involves pausing of transcription by TTF1, and the dissociation of the transcription complex, releasing pre-rRNA and RNA polymerase I from the template. PTRF is required for dissociation of the ternary transcription complex.Specifications
| PTRF | |
| Polyclonal | |
| Unconjugated | |
| CAVIN1 | |
| Polymerase I and transcript release factor; PTRF; FKSG13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 284119 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q6NZI2 | |
| CAVIN1 | |
| Between 150 and 200 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.